siRNA Details
| Gene Symbol: Tmprss11g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177162.4 | NP_796136.2 | 5 | 544 | ||
| GenBank mRNA | AK036981.1 | - | 5 | 545 | ||
| AK037029.1 | - | 1 | 42 | |||
| BC118611.1 | - | 5 | 522 | 40107184 | ||
| BC118660.1 | - | 5 | 521 | 40107187 | ||
| CK627573.1 | - | 5 | 602 | |||

Tmprss11g
Genomic Map