siRNA Details
| Gene Symbol: Tmprss11g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177162.4 | NP_796136.2 | 10 | 1871 | ||
| GenBank mRNA | AK036981.1 | - | 10 | 1874 | ||
| AK037029.1 | - | 6 | 1370 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmprss11g| Gene Symbol: Tmprss11g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177162.4 | NP_796136.2 | 10 | 1871 | ||
| GenBank mRNA | AK036981.1 | - | 10 | 1874 | ||
| AK037029.1 | - | 6 | 1370 | |||