siRNA Details
| Gene Symbol: TSSC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF125568.1 | - | 2 | 1415 | ||
| BC050616.1 | - | 4 | 1533 | 5744058 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TSSC4| Gene Symbol: TSSC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF125568.1 | - | 2 | 1415 | ||
| BC050616.1 | - | 4 | 1533 | 5744058 | ||