siRNA Details
| Gene Symbol: Ubn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177185.4 | NP_796159.3 | 4 | 883 | ||
| GenBank mRNA | AK031359.1 | - | 5 | 851 | ||
| AK051609.1 | - | 4 | 250 | |||
| BC043095.1 | - | 131 | ||||
| BC051458.1 | - | 4 | 858 | 6468017 | ||

Ubn2
Genomic Map