siRNA Details
| Gene Symbol: Ubn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177185.4 | NP_796159.3 | 4, 5 | 1006 | ||
| GenBank mRNA | AK051609.1 | - | 4, 5 | 373 | ||
| BC043095.1 | - | 254 | ||||
| BC051458.1 | - | 4, 5 | 981 | 6468017 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ubn2| Gene Symbol: Ubn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177185.4 | NP_796159.3 | 4, 5 | 1006 | ||
| GenBank mRNA | AK051609.1 | - | 4, 5 | 373 | ||
| BC043095.1 | - | 254 | ||||
| BC051458.1 | - | 4, 5 | 981 | 6468017 | ||