siRNA Details
| Gene Symbol: Slc35e2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177186.4 | NP_796160.1 | 2, 3 | 804 | ||
| GenBank mRNA | AK048716.1 | - | 2, 3 | 673 | ||
| AK080215.1 | - | 2, 3 | 823 | |||
| AK145065.1 | - | 3, 4 | 727 | |||
| AK157354.1 | - | 2, 3 | 700 | |||
| BC058728.1 | - | 2, 3 | 804 | 6407259 | ||

Slc35e2
Genomic Map