siRNA Details
| Gene Symbol: Ccdc169 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290138.1 | NP_001277067.1 | 8 | 1276 | ||
| NM_001290141.1 | NP_001277070.1 | 8 | 751 | |||
| NM_177203.3 | NP_796177.2 | 8 | 655 | |||
| GenBank mRNA | AK042903.1 | - | 8 | 1278 | ||
| BC150827.1 | - | 7 | 483 | |||
| BC171976.1 | - | 8 | 510 | |||
| BC171978.1 | - | 9 | 544 | |||
| BC171981.1 | - | 7 | 483 | |||

Ccdc169
Genomic Map