siRNA Details
| Gene Symbol: Zfp629 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177226.5 | NP_796200.1 | 3 | 3322 | ||
| GenBank mRNA | AK031161.1 | - | 3 | 3031 | ||
| AK034484.1 | - | 3 | 2994 | |||
| AK172933.1 | - | 4 | 3052 | |||
| BC035547.1 | - | 3 | 2964 | 4481313 | ||
| BC067053.1 | - | 3 | 3322 | 30543652 | ||

Zfp629
Genomic Map