siRNA Details
| Gene Symbol: Mysm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177239.2 | NP_796213.2 | 8 | 828 | ||
| GenBank mRNA | AK043802.1 | - | 8 | 828 | ||
| AK081684.1 | - | 8 | 803 | |||
| AK173300.1 | - | 5 | 570 | |||
| BC150946.1 | - | 8 | 824 | 9087857 | ||
| BC151172.1 | - | 8 | 824 | 9088061 | ||

Mysm1
Genomic Map