siRNA Details
| Gene Symbol: 9230112E08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK030308.1 | - | 249 | |||
| AK033807.1 | - | 431 | ||||
| BC057100.1 | - | 355 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

9230112E08Rik| Gene Symbol: 9230112E08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK030308.1 | - | 249 | |||
| AK033807.1 | - | 431 | ||||
| BC057100.1 | - | 355 | ||||