siRNA Details
Gene Symbol: 9230112E08Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK030308.1 | - | 700 | |||
AK033807.1 | - | 881 | ||||
BC057100.1 | - | 805 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 9230112E08Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK030308.1 | - | 700 | |||
AK033807.1 | - | 881 | ||||
BC057100.1 | - | 805 |