siRNA Details
| Gene Symbol: Ifitm10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177265.4 | NP_796239.1 | 4 | 1484 | ||
| GenBank mRNA | AK031961.1 | - | 3 | 1043 | ||
| AK045822.1 | - | 4 | 1485 | |||
| AK076361.1 | - | 2 | 957 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ifitm10| Gene Symbol: Ifitm10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177265.4 | NP_796239.1 | 4 | 1484 | ||
| GenBank mRNA | AK031961.1 | - | 3 | 1043 | ||
| AK045822.1 | - | 4 | 1485 | |||
| AK076361.1 | - | 2 | 957 | |||