siRNA Details
Gene Symbol: KIF20A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005733.2 | NP_005724.1 | 16 | 2574 | ||
GenBank mRNA | AF070672.1 | - | 15 | 2105 | ||
AF153329.1 | - | 16 | 2112 | |||
AK024804.1 | - | 5 | 631 | |||
AK025790.1 | - | 16 | 2133 | |||
AK296879.1 | - | 17 | 2088 | |||
BC012999.2 | - | 16 | 2112 | 3604291 |
