siRNA Details
| Gene Symbol: Spata33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK028607.1 | - | 1 | 2536 | ||
| AK049895.1 | - | 2 | 1423 | |||
| AK053679.1 | - | 2 | 1223 | |||
| BC059919.1 | - | 1 | 2484 | 6827099 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Spata33| Gene Symbol: Spata33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK028607.1 | - | 1 | 2536 | ||
| AK049895.1 | - | 2 | 1423 | |||
| AK053679.1 | - | 2 | 1223 | |||
| BC059919.1 | - | 1 | 2484 | 6827099 | ||