siRNA Details
| Gene Symbol: D830039M14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK028369.1 | - | 1 | 1463 | ||
| AK039329.1 | - | 1 | 1208 | |||
 Back To Top
 Back To TopPlease enter the information below and press OK to send your cart to Core Services for purchase.

 D830039M14Rik
D830039M14Rik| Gene Symbol: D830039M14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK028369.1 | - | 1 | 1463 | ||
| AK039329.1 | - | 1 | 1208 | |||
 Back To Top
 Back To Top