siRNA Details
| Gene Symbol: 6330408A02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177312.4 | NP_796286.2 | 4 | 894 | ||
| GenBank mRNA | AK018141.1 | - | 4 | 654 | ||
| AK160064.1 | - | 3 | 1533 | |||
| BC075681.1 | - | 3 | 868 | 30608389 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

6330408A02Rik| Gene Symbol: 6330408A02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177312.4 | NP_796286.2 | 4 | 894 | ||
| GenBank mRNA | AK018141.1 | - | 4 | 654 | ||
| AK160064.1 | - | 3 | 1533 | |||
| BC075681.1 | - | 3 | 868 | 30608389 | ||