siRNA Details
| Gene Symbol: Fam166b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001162381.1 | NP_001155853.1 | 5 | 853 | ||
| NM_177377.5 | NP_796351.2 | 4 | 930 | |||
| GenBank mRNA | AK029435.1 | - | 4 | 931 | ||
| AK053208.1 | - | 4 | 931 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam166b| Gene Symbol: Fam166b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001162381.1 | NP_001155853.1 | 5 | 853 | ||
| NM_177377.5 | NP_796351.2 | 4 | 930 | |||
| GenBank mRNA | AK029435.1 | - | 4 | 931 | ||
| AK053208.1 | - | 4 | 931 | |||