siRNA Details
| Gene Symbol: Ttc16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290563.1 | NP_001277492.1 | 12 | 1847 | ||
| NM_001290564.1 | NP_001277493.1 | 12 | 1902 | |||
| NM_001290566.1 | NP_001277495.1 | 11 | 1600 | |||
| NM_001290567.1 | NP_001277496.1 | 12 | 1648 | |||
| NM_177384.3 | NP_796358.2 | 12 | 1847 | |||
| GenBank mRNA | AK028040.1 | - | 12 | 1902 | ||
| AK089384.1 | - | 9 | 2703 | |||
| AK133273.1 | - | 12 | 1625 | |||
| AK160435.1 | - | 12 | 1598 | |||
| BC140991.1 | - | 11 | 1591 | 9055603 | ||
| BC145064.1 | - | 12 | 1639 | 9053612 | ||

Ttc16
Genomic Map