siRNA Details
| Gene Symbol: Cntln | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177385.4 | NP_796359.1 | 5 | 1695 | ||
| GenBank mRNA | AK031604.1 | - | 5 | 1601 | ||
| AK165111.1 | - | 4 | 1168 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cntln| Gene Symbol: Cntln | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177385.4 | NP_796359.1 | 5 | 1695 | ||
| GenBank mRNA | AK031604.1 | - | 5 | 1601 | ||
| AK165111.1 | - | 4 | 1168 | |||