siRNA Details
| Gene Symbol: Tmem220 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291042.1 | NP_001277971.1 | 6 | 923 | ||
| NM_001291043.1 | NP_001277972.1 | 6 | 919 | |||
| NM_177392.3 | NP_796366.1 | 7 | 995 | |||
| GenBank mRNA | AK043094.1 | - | 7 | 996 | ||
| AK078505.1 | - | 6 | 922 | |||
| BC110628.1 | - | 6 | 923 | 40052084 | ||
| BC110629.1 | - | 6 | 919 | 40052085 | ||

Tmem220
Genomic Map