siRNA Details
| Gene Symbol: Slfnl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177570.3 | NP_808238.1 | 3 | 1072 | ||
| GenBank mRNA | AK077106.1 | - | 3 | 1073 | ||
| BC117545.1 | - | 2 | 820 | 40105640 | ||
| BC118052.1 | - | 2 | 823 | 40105639 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slfnl1| Gene Symbol: Slfnl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177570.3 | NP_808238.1 | 3 | 1072 | ||
| GenBank mRNA | AK077106.1 | - | 3 | 1073 | ||
| BC117545.1 | - | 2 | 820 | 40105640 | ||
| BC118052.1 | - | 2 | 823 | 40105639 | ||