siRNA Details
| Gene Symbol: Skint3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177578.4 | NP_808246.1 | 8 | 2559 | ||
| GenBank mRNA | AK040383.1 | - | 8 | 2561 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Skint3| Gene Symbol: Skint3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177578.4 | NP_808246.1 | 8 | 2559 | ||
| GenBank mRNA | AK040383.1 | - | 8 | 2561 | ||