siRNA Details
Gene Symbol: Gm9871 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK027905.1 | - | 6 | 1230 | ||
BC125571.1 | - | 5 | 863 | |||
BC145947.1 | - | 5 | 863 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm9871 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK027905.1 | - | 6 | 1230 | ||
BC125571.1 | - | 5 | 863 | |||
BC145947.1 | - | 5 | 863 |