siRNA Details
| Gene Symbol: Cdnf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177647.4 | NP_808315.1 | 2 | 205 | ||
| GenBank mRNA | AK034009.1 | - | 2 | 207 | ||
| BC110560.2 | - | 2 | 193 | 40049634 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cdnf| Gene Symbol: Cdnf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177647.4 | NP_808315.1 | 2 | 205 | ||
| GenBank mRNA | AK034009.1 | - | 2 | 207 | ||
| BC110560.2 | - | 2 | 193 | 40049634 | ||