siRNA Details
| Gene Symbol: D630003M21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177657.5 | NP_808325.2 | 14 | 3839 | ||
| GenBank mRNA | AK052605.1 | - | 14 | 3838 | ||
| BB496563.2 | - | 1 | 574 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

D630003M21Rik| Gene Symbol: D630003M21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177657.5 | NP_808325.2 | 14 | 3839 | ||
| GenBank mRNA | AK052605.1 | - | 14 | 3838 | ||
| BB496563.2 | - | 1 | 574 | |||