siRNA Details
Gene Symbol: Epha10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177671.5 | NP_808339.2 | 3 | 1082 | ||
GenBank mRNA | AK039700.1 | - | 3 | 1045 | ||
AK143168.1 | - | 1967 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Epha10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177671.5 | NP_808339.2 | 3 | 1082 | ||
GenBank mRNA | AK039700.1 | - | 3 | 1045 | ||
AK143168.1 | - | 1967 |