siRNA Details
| Gene Symbol: Crebl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177687.3 | NP_808355.1 | 2 | 321 | ||
| GenBank mRNA | AK045467.1 | - | 2 | 322 | ||
| AK195782.1 | - | 2 | 95 | |||
| BC109358.1 | - | 2 | 193 | 40057562 | ||
| BC109359.1 | - | 2 | 209 | 40057564 | ||

Crebl2
Genomic Map