siRNA Details
| Gene Symbol: H2afj | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177688.4 | NP_808356.1 | 1 | 532 | ||
| GenBank mRNA | AK053755.1 | - | 1 | 457 | ||
| BC024397.1 | - | 1 | 492 | 5055276 | ||
| BC099606.1 | - | 1 | 514 | 1383389 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

H2afj| Gene Symbol: H2afj | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177688.4 | NP_808356.1 | 1 | 532 | ||
| GenBank mRNA | AK053755.1 | - | 1 | 457 | ||
| BC024397.1 | - | 1 | 492 | 5055276 | ||
| BC099606.1 | - | 1 | 514 | 1383389 | ||