siRNA Details
| Gene Symbol: Vwa3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177697.3 | NP_808365.2 | 18 | 2096 | ||
| GenBank mRNA | AK086942.1 | - | 17 | 1694 | ||
| AK132732.1 | - | 18 | 2097 | |||
| AK136898.1 | - | 17 | 1695 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vwa3a| Gene Symbol: Vwa3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177697.3 | NP_808365.2 | 18 | 2096 | ||
| GenBank mRNA | AK086942.1 | - | 17 | 1694 | ||
| AK132732.1 | - | 18 | 2097 | |||
| AK136898.1 | - | 17 | 1695 | |||