siRNA Details
| Gene Symbol: Psd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027626.1 | NP_081902.1 | 4 | 641 | ||
| NM_030263.5 | NP_084539.2 | 9 | 983 | |||
| NM_177698.4 | NP_808366.2 | 12 | 2747 | |||
| GenBank mRNA | AB220685.1 | - | 12 | 2747 | ||
| AK031477.1 | - | 4 | 584 | |||
| AK122405.1 | - | 3 | 297 | |||
| AK147606.1 | - | 4 | 641 | |||
| AK161498.1 | - | 9 | 969 | |||
| BC052365.1 | - | 9 | 948 | |||
| BC131913.1 | - | 4 | 553 | |||
| BC145641.1 | - | 4 | 553 | |||

Psd3
Genomic Map