siRNA Details
| Gene Symbol: Mcmdc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177722.3 | NP_808390.2 | 5, 6 | 586 | ||
| GenBank mRNA | AK031395.1 | - | 5, 6 | 544 | ||
| CA978963.1 | - | 3, 4 | 341 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mcmdc2| Gene Symbol: Mcmdc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177722.3 | NP_808390.2 | 5, 6 | 586 | ||
| GenBank mRNA | AK031395.1 | - | 5, 6 | 544 | ||
| CA978963.1 | - | 3, 4 | 341 | |||