siRNA Details
| Gene Symbol: C130073F10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AA607315.1 | - | 1, 2 | 32 | ||
| AK081744.1 | - | 2, 3 | 387 | |||
| BB374054.2 | - | 1, 2 | 99 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C130073F10Rik| Gene Symbol: C130073F10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AA607315.1 | - | 1, 2 | 32 | ||
| AK081744.1 | - | 2, 3 | 387 | |||
| BB374054.2 | - | 1, 2 | 99 | |||