siRNA Details
| Gene Symbol: Tmem130 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177735.4 | NP_808403.2 | 4 | 714 | ||
| GenBank mRNA | AK081543.1 | - | 1 | 81 | ||
| BC067004.1 | - | 4 | 714 | 6416222 | ||
| BC127616.1 | - | 4 | 691 | 40093666 | ||
| BC127617.1 | - | 4 | 691 | 40093667 | ||

Tmem130
Genomic Map