siRNA Details
| Gene Symbol: Fam198a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199927.1 | NP_001186856.1 | 4 | 1616 | ||
| NM_177743.5 | NP_808411.1 | 4 | 1547 | |||
| GenBank mRNA | AK050223.1 | - | 4 | 1535 | ||
| AK134876.1 | - | 4 | 1602 | |||
| AK139038.1 | - | 4 | 1617 | |||
| AK158406.1 | - | 4 | 1549 | |||
| BC113767.1 | - | 3 | 1396 | 40058404 | ||

Fam198a
Genomic Map