siRNA Details
| Gene Symbol: Slc35e1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177766.3 | NP_808434.2 | 5, 6 | 1040 | ||
| GenBank mRNA | AK031600.1 | - | 5, 6 | 788 | ||
| AK200930.1 | - | 2, 3 | 303 | |||
| AK203664.1 | - | 1, 2 | 306 | |||
| BC115553.1 | - | 5, 6 | 699 | 40101801 | ||
| BC115554.1 | - | 5, 6 | 699 | 40101802 | ||

Slc35e1
Genomic Map