siRNA Details
| Gene Symbol: Slc15a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177787.4 | NP_808455.2 | 5 | 972 | ||
| GenBank mRNA | AK036413.1 | - | 4 | 755 | ||
| BC026899.1 | - | 2 | 50 | |||
| BC039761.1 | - | 2 | 61 | |||
| BC141332.1 | - | 4 | 691 | 9055944 | ||
| BC141334.1 | - | 4 | 691 | 9055946 | ||

Slc15a5
Genomic Map