siRNA Details
| Gene Symbol: DDX39A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005804.3 | NP_005795.2 | 2 | 231 | ||
| NR_046366.2 | 2 | 231 | ||||
| XM_006722606.3 | XP_006722669.1 | 2 | 171 | |||
| XM_011527620.1 | XP_011525922.1 | 2 | 250 | |||
| XM_011527621.2 | XP_011525923.1 | 1 | 117 | |||
| GenBank mRNA | AB254849.1 | - | 1 | 113 | ||
| AK293911.1 | - | 2 | 189 | |||
| AK301847.1 | - | 3 | 314 | |||
| BC001009.2 | - | 2 | 168 | 2820926 | ||
| BC032128.2 | - | 2 | 177 | 5017293 | ||
| DA432925.1 | - | 2 | 232 | |||
| DB032777.1 | - | 3 | 324 | |||
| U90426.1 | - | 2 | 208 | |||

DDX39A
Genomic Map