siRNA Details
| Gene Symbol: Lrrc16a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026825.3 | NP_081101.3 | 3, 4 | 387 | ||
| GenBank mRNA | AK051570.1 | - | 3, 4 | 387 | ||
| AY437876.1 | - | 3, 4 | 392 | |||
| BC150819.1 | - | 3, 4 | 313 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lrrc16a| Gene Symbol: Lrrc16a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026825.3 | NP_081101.3 | 3, 4 | 387 | ||
| GenBank mRNA | AK051570.1 | - | 3, 4 | 387 | ||
| AY437876.1 | - | 3, 4 | 392 | |||
| BC150819.1 | - | 3, 4 | 313 | |||