siRNA Details
| Gene Symbol: PRG4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127709.2 | NP_001121181.2 | 3 | 168 | ||
| NM_001303232.1 | NP_001290161.1 | 3 | 168 | |||
| NM_005807.4 | NP_005798.3 | 3 | 168 | |||
| XM_017000002.1 | XP_016855491.1 | 2 | 464 | |||
| GenBank mRNA | AK297718.1 | - | 2 | 162 | ||
| AK301343.1 | - | 3 | 163 | |||
| DA995681.1 | - | 3 | 168 | |||
| DC404857.1 | - | 2 | 162 | |||
| U70136.1 | - | 2 | 150 | |||

PRG4
Genomic Map