siRNA Details
| Gene Symbol: Zfp459 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177811.4 | NP_808479.1 | 4 | 317 | ||
| GenBank mRNA | AK036923.1 | - | 4 | 318 | ||
| BC100389.1 | - | 4 | 283 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp459| Gene Symbol: Zfp459 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177811.4 | NP_808479.1 | 4 | 317 | ||
| GenBank mRNA | AK036923.1 | - | 4 | 318 | ||
| BC100389.1 | - | 4 | 283 | |||