siRNA Details
| Gene Symbol: Fam135b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177819.3 | NP_808487.2 | 7, 8 | 866 | ||
| GenBank mRNA | AK043571.1 | - | 7, 8 | 867 | ||
| BC125645.1 | - | 7, 8 | 741 | |||
| BC125647.1 | - | 7, 8 | 741 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam135b| Gene Symbol: Fam135b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177819.3 | NP_808487.2 | 7, 8 | 866 | ||
| GenBank mRNA | AK043571.1 | - | 7, 8 | 867 | ||
| BC125645.1 | - | 7, 8 | 741 | |||
| BC125647.1 | - | 7, 8 | 741 | |||