siRNA Details
| Gene Symbol: GDF11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005811.3 | NP_005802.1 | 2 | 552 | ||
| XM_006719194.3 | XP_006719257.1 | 2 | 554 | |||
| GenBank mRNA | AF028333.1 | - | 2 | 433 | ||
| AF100907.1 | - | 2 | 553 | |||
| KC561773.1 | - | 1 | 174 | |||

GDF11
Genomic Map