siRNA Details
| Gene Symbol: Fam163a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177838.3 | NP_808506.1 | 1524 | |||
| GenBank mRNA | AK039197.1 | - | 1525 | |||
| BC116972.1 | - | 2 | 1193 | 40126291 | ||
| BC116974.1 | - | 2 | 1193 | 40126293 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam163a| Gene Symbol: Fam163a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177838.3 | NP_808506.1 | 1524 | |||
| GenBank mRNA | AK039197.1 | - | 1525 | |||
| BC116972.1 | - | 2 | 1193 | 40126291 | ||
| BC116974.1 | - | 2 | 1193 | 40126293 | ||