siRNA Details
| Gene Symbol: Bpi | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177850.3 | NP_808518.1 | 2 | 287 | ||
| GenBank mRNA | AK033770.1 | - | 2 | 289 | ||
| AY363993.1 | - | 2 | 285 | |||
| AY648037.1 | - | 2 | 287 | |||
| AY853179.1 | - | 2 | 272 | |||
| BC118049.1 | - | 2 | 281 | 40106697 | ||
| BC118504.1 | - | 2 | 281 | 40106694 | ||

Bpi
Genomic Map