siRNA Details
| Gene Symbol: Tmem67 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177861.4 | NP_808529.2 | 5, 6 | 592 | ||
| GenBank mRNA | AK017238.1 | - | 5, 6 | 564 | ||
| AK045429.1 | - | 5, 6 | 593 | |||
| AK084054.1 | - | 1373 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem67| Gene Symbol: Tmem67 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177861.4 | NP_808529.2 | 5, 6 | 592 | ||
| GenBank mRNA | AK017238.1 | - | 5, 6 | 564 | ||
| AK045429.1 | - | 5, 6 | 593 | |||
| AK084054.1 | - | 1373 | ||||