siRNA Details
| Gene Symbol: Frem1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001198811.1 | NP_001185740.1 | 25 | 5209 | ||
| NM_177863.4 | NP_808531.2 | 24 | 4999 | |||
| GenBank mRNA | AB160986.1 | - | 25 | 5209 | ||
| AJ616838.1 | - | 24 | 4999 | |||
| AK052629.1 | - | 8 | 1513 | |||
| BC138427.1 | - | 24 | 4910 | 8861447 | ||
| BC145274.1 | - | 24 | 4963 | |||

Frem1
Genomic Map