siRNA Details
| Gene Symbol: C130026L21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK047985.1 | - | 1024 | |||
| AK160721.1 | - | 4 | 1183 | |||
| BC094571.1 | - | 3 | 859 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | NM_177875.1 | - | 1024 | Chr. - - [-] on Build | ||

C130026L21Rik
Genomic Map