siRNA Details
| Gene Symbol: Fam188b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177883.4 | NP_808551.2 | 11 | 2087 | ||
| GenBank mRNA | AK082855.1 | - | 11 | 2088 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam188b| Gene Symbol: Fam188b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177883.4 | NP_808551.2 | 11 | 2087 | ||
| GenBank mRNA | AK082855.1 | - | 11 | 2088 | ||