siRNA Details
| Gene Symbol: Gcfc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177884.3 | NP_808552.2 | 5 | 771 | ||
| GenBank mRNA | AK038369.1 | - | 6 | 844 | ||
| AK050789.1 | - | 5 | 713 | |||
| AK077263.1 | - | 5 | 761 | |||
| AK078171.1 | - | 5 | 762 | |||
| AK136387.1 | - | 3 | 373 | |||

Gcfc2
Genomic Map