siRNA Details
| Gene Symbol: Gcfc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177884.3 | NP_808552.2 | 7, 8 | 1147 | ||
| GenBank mRNA | AK038369.1 | - | 8, 9 | 1220 | ||
| AK050789.1 | - | 7, 8 | 1089 | |||
| AK077263.1 | - | 7, 8 | 1137 | |||
| AK078171.1 | - | 7, 8 | 1138 | |||
| AK136387.1 | - | 5, 6 | 749 | |||

Gcfc2
Genomic Map